Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616097 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C11orf98 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C11orf98 - chromosome 11 open reading frame 98 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LBHD1 - LBH domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024099.3 | 740 | Intron | NP_077004.2 |
METTL12 - methyltransferase like 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001043229.1 | 740 | Missense Mutation | CGG,TGG | R,W 38 | NP_001036694.1 | |
XM_005274232.4 | 740 | Missense Mutation | CGG,TGG | R,W 161 | XP_005274289.1 |
SNORA57 - small nucleolar RNA, H/ACA box 57 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UQCC3 - ubiquinol-cytochrome c reductase complex assembly factor 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |