Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCCAAGATGTCATTGTGGTTTCAA[A/G]TCTAAAAAAAAGTTTATATATGATT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609342 | ||||||||||||||||||||
Literature Links: |
GIF PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GIF - gastric intrinsic factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005142.2 | 1109 | Missense Mutation | CTT,TTT | L,F 359 | NP_005133.2 | |
XM_011544939.2 | 1109 | Missense Mutation | CTT,TTT | L,F 345 | XP_011543241.1 |