Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTTGGAGTTGTTGACTGGCATGGC[A/G]GAGATCTTTGGCACAGAGCTGACCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608757 MIM: 609725 MIM: 614586 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CLP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CLP1 - cleavage and polyadenylation factor I subunit 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142597.1 | 265 | Silent Mutation | GCA,GCG | A,A 42 | NP_001136069.1 | |
NM_006831.2 | 265 | Silent Mutation | GCA,GCG | A,A 42 | NP_006822.1 |
YPEL4 - yippee like 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZDHHC5 - zinc finger DHHC-type containing 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |