Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTATATTTCCACAACAAACCTATC[C/T]TTTGGTGCTATGAATGTGTTTACAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608401 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR6503 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MIR6503 - microRNA 6503 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MS4A4E - membrane spanning 4-domains A4E | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_011545416.2 | Intron | XP_011543718.1 | ||||
XM_011545417.2 | Intron | XP_011543719.1 | ||||
XM_011545418.2 | Intron | XP_011543720.1 | ||||
XM_011545419.2 | Intron | XP_011543721.1 | ||||
XM_017018641.1 | Intron | XP_016874130.1 |