Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTGACTTGCCTAAGATCACAGAAT[A/C]ATTATGTGGCAGGAACAACATTCAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615944 MIM: 611117 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C2CD3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C2CD3 - C2 calcium dependent domain containing 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PPME1 - protein phosphatase methylesterase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271593.1 | Intron | NP_001258522.1 | ||||
NM_016147.2 | Intron | NP_057231.1 | ||||
XM_017017913.1 | Intron | XP_016873402.1 |