Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCCTCATTCTTACTTCCACTCTGA[A/G]TATCTGTGGGATGGAAAAGACACAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 276903 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GDPD4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GDPD4 - glycerophosphodiester phosphodiesterase domain containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_182833.1 | 2006 | Missense Mutation | ACT,ATT | T,I 510 | NP_878253.1 | |
XM_011544826.1 | 2006 | Missense Mutation | ACT,ATT | T,I 312 | XP_011543128.1 | |
XM_011544833.1 | 2006 | Intron | XP_011543135.1 | |||
XM_011544834.1 | 2006 | Missense Mutation | ACT,ATT | T,I 536 | XP_011543136.1 | |
XM_017017348.1 | 2006 | Intron | XP_016872837.1 | |||
XM_017017349.1 | 2006 | Intron | XP_016872838.1 | |||
XM_017017350.1 | 2006 | Intron | XP_016872839.1 | |||
XM_017017351.1 | 2006 | Intron | XP_016872840.1 |
MYO7A - myosin VIIA | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |