Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCTCCTTGACCCTGAAACGTGCAG[C/T]AAGTAGATAACCTCCTGATCCCTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC100133315 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC100133315 - transient receptor potential cation channel, subfamily C, member 2-like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RNF121 - ring finger protein 121 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300926.1 | 188 | UTR 5 | NP_001287855.1 | |||
NM_018320.4 | 188 | UTR 5 | NP_060790.2 | |||
XM_006718629.1 | 188 | UTR 5 | XP_006718692.1 | |||
XM_006718630.1 | 188 | Intron | XP_006718693.1 | |||
XM_006718632.1 | 188 | UTR 5 | XP_006718695.1 | |||
XM_011545150.1 | 188 | Intron | XP_011543452.1 |