Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTAAGAGAAACTCTGTAAGTTAAA[A/G]AAGTGCCACATAATTAATTTCTTAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604353 MIM: 611636 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHORDC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CHORDC1 - cysteine and histidine rich domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001144073.1 | Intron | NP_001137545.1 | ||||
NM_012124.2 | Intron | NP_036256.2 | ||||
XM_017017540.1 | Intron | XP_016873029.1 | ||||
XM_017017541.1 | Intron | XP_016873030.1 |
NAALAD2 - N-acetylated alpha-linked acidic dipeptidase 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |