Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGGGGGCCATGGCCTAAAAGGGG[A/G]CTAGTTCCGGAAAGCCCAGGCCCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 114220 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CAPN1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CAPN1 - calpain 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001198868.1 | Intron | NP_001185797.1 | ||||
NM_001198869.1 | Intron | NP_001185798.1 | ||||
NM_005186.3 | Intron | NP_005177.2 | ||||
XM_006718698.1 | Intron | XP_006718761.1 | ||||
XM_011545292.1 | Intron | XP_011543594.1 |