Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGAACAGGCCATGCCAGGGGCTGG[C/G]CCTGGTGTGCCCCAGCTGGGAAACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606547 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MS4A4A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MS4A4A - membrane spanning 4-domains A4A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243266.1 | Intron | NP_001230195.1 | ||||
NM_024021.3 | Intron | NP_076926.2 | ||||
NM_148975.2 | Intron | NP_683876.1 | ||||
XM_017017909.1 | Intron | XP_016873398.1 |