Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACATTTCTCAGCACTGGACTTTAA[A/G]CTACTCTGAGAATCCTTCCCTCCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609651 MIM: 601644 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GPHA2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GPHA2 - glycoprotein hormone alpha 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MAJIN - membrane anchored junction protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001037225.2 | Intron | NP_001032302.1 | ||||
NM_001300803.1 | Intron | NP_001287732.1 | ||||
NM_001318808.1 | Intron | NP_001305737.1 | ||||
XM_005273918.2 | Intron | XP_005273975.1 | ||||
XM_006718502.2 | Intron | XP_006718565.1 | ||||
XM_006718503.3 | Intron | XP_006718566.1 | ||||
XM_006718504.2 | Intron | XP_006718567.1 | ||||
XM_006718505.2 | Intron | XP_006718568.1 | ||||
XM_011544940.2 | Intron | XP_011543242.1 | ||||
XM_011544941.2 | Intron | XP_011543243.1 |
PPP2R5B - protein phosphatase 2 regulatory subunit B'beta | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |