Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGGAATAATCTTAAAATCATATTA[G/T]AGGTTTTTTAATTTTTTTGAAAAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604102 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GRM5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GRM5 - glutamate metabotropic receptor 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000842.4 | 8043 | UTR 3 | NP_000833.1 | |||
NM_001143831.2 | 8043 | UTR 3 | NP_001137303.1 | |||
XM_006718828.3 | 8043 | UTR 3 | XP_006718891.1 | |||
XM_011542792.1 | 8043 | UTR 3 | XP_011541094.1 | |||
XM_017017627.1 | 8043 | UTR 3 | XP_016873116.1 |
GRM5-AS1 - GRM5 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |