Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTAAATACTTAATTGAACTTTTTC[C/T]CTAGGATCTAACATAGAATAAGAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 176785 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DDIAS PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DDIAS - DNA damage induced apoptosis suppressor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145018.3 | Intron | NP_659455.3 | ||||
XM_011544835.1 | Intron | XP_011543137.1 | ||||
XM_011544836.2 | Intron | XP_011543138.1 | ||||
XM_011544837.1 | Intron | XP_011543139.1 | ||||
XM_011544838.2 | Intron | XP_011543140.1 | ||||
XM_017017352.1 | Intron | XP_016872841.1 | ||||
XM_017017353.1 | Intron | XP_016872842.1 |
PRCP - prolylcarboxypeptidase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |