Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGTAAAGCCAAATAACATGGCTGG[A/G]ATTCTTTAGACGGTGCAGGTTGTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600454 MIM: 600455 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RPS3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RPS3 - ribosomal protein S3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001005.4 | Intron | NP_000996.2 | ||||
NM_001256802.1 | Intron | NP_001243731.1 | ||||
NM_001260506.1 | Intron | NP_001247435.1 | ||||
NM_001260507.1 | Intron | NP_001247436.1 |
SNORD15A - small nucleolar RNA, C/D box 15A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORD15B - small nucleolar RNA, C/D box 15B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |