Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTTCTCTAGCTAGTAAATGACAGT[A/G]TCTTAATGGGCTTTGGATTTAATGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607951 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CEP57 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CEP57 - centrosomal protein 57 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243776.1 | 684 | Intron | NP_001230705.1 | |||
NM_001243777.1 | 684 | Intron | NP_001230706.1 | |||
NM_014679.4 | 684 | Intron | NP_055494.2 | |||
XM_006718945.2 | 684 | Intron | XP_006719008.1 | |||
XM_006718946.2 | 684 | Intron | XP_006719009.1 | |||
XM_017018592.1 | 684 | UTR 5 | XP_016874081.1 | |||
XM_017018593.1 | 684 | Intron | XP_016874082.1 | |||
XM_017018594.1 | 684 | Intron | XP_016874083.1 |
FAM76B - family with sequence similarity 76 member B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |