Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGCCCTGAATAACGAATCTCAGTT[C/T]TTCTTAAGGTCCTCCTCCTCTGAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611474 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HRASLS5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HRASLS5 - HRAS like suppressor family member 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001146728.1 | 1340 | UTR 3 | NP_001140200.1 | |||
NM_001146729.1 | 1340 | UTR 3 | NP_001140201.1 | |||
NM_054108.3 | 1340 | UTR 3 | NP_473449.1 | |||
XM_011544751.2 | 1340 | UTR 3 | XP_011543053.1 | |||
XM_011544752.2 | 1340 | UTR 3 | XP_011543054.1 | |||
XM_011544753.1 | 1340 | UTR 3 | XP_011543055.1 | |||
XM_017017172.1 | 1340 | UTR 3 | XP_016872661.1 |