Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATATATGCACAAAAAACAATTCAA[A/G]TAAGAGTTCAGAGTAAAAAAAAGTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 118491 MIM: 604592 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHKA PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CHKA - choline kinase alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001277.2 | 3999 | UTR 3 | NP_001268.2 | |||
NM_212469.1 | 3999 | UTR 3 | NP_997634.1 | |||
XM_017017147.1 | 3999 | Intron | XP_016872636.1 | |||
XM_017017148.1 | 3999 | UTR 3 | XP_016872637.1 |
MIR6753 - microRNA 6753 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TCIRG1 - T-cell immune regulator 1, ATPase H+ transporting V0 subunit a3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006019.3 | 3999 | Intron | NP_006010.2 | |||
NM_006053.3 | 3999 | Intron | NP_006044.1 | |||
XM_005273709.3 | 3999 | Intron | XP_005273766.1 | |||
XM_011544726.2 | 3999 | Intron | XP_011543028.1 | |||
XM_017017089.1 | 3999 | Intron | XP_016872578.1 | |||
XM_017017090.1 | 3999 | Intron | XP_016872579.1 |