Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAAAAAGGGAAAAAATTACACCTG[C/T]ACAACCAATAGACATACAAAGTTAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607951 MIM: 603557 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CEP57 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CEP57 - centrosomal protein 57 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243776.1 | 4175 | Intron | NP_001230705.1 | |||
NM_001243777.1 | 4175 | Intron | NP_001230706.1 | |||
NM_014679.4 | 4175 | Intron | NP_055494.2 | |||
XM_006718945.2 | 4175 | Intron | XP_006719008.1 | |||
XM_006718946.2 | 4175 | Intron | XP_006719009.1 | |||
XM_017018592.1 | 4175 | Intron | XP_016874081.1 | |||
XM_017018593.1 | 4175 | Intron | XP_016874082.1 | |||
XM_017018594.1 | 4175 | Intron | XP_016874083.1 |
MTMR2 - myotubularin related protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243571.1 | 4175 | UTR 3 | NP_001230500.1 | |||
NM_016156.5 | 4175 | UTR 3 | NP_057240.3 | |||
NM_201278.2 | 4175 | UTR 3 | NP_958435.1 | |||
NM_201281.2 | 4175 | UTR 3 | NP_958438.1 | |||
XM_005274374.2 | 4175 | UTR 3 | XP_005274431.1 | |||
XM_005274375.2 | 4175 | UTR 3 | XP_005274432.1 | |||
XM_006718934.2 | 4175 | UTR 3 | XP_006718997.1 | |||
XM_006718935.2 | 4175 | UTR 3 | XP_006718998.1 | |||
XM_006718936.3 | 4175 | UTR 3 | XP_006718999.1 | |||
XM_011543058.2 | 4175 | UTR 3 | XP_011541360.1 | |||
XM_011543059.2 | 4175 | UTR 3 | XP_011541361.1 | |||
XM_017018517.1 | 4175 | UTR 3 | XP_016874006.1 | |||
XM_017018518.1 | 4175 | UTR 3 | XP_016874007.1 |