Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACTGTCTGTTTGTTGAAACTCTGG[A/G]GCGAAGGCACTGTGGCCTGGTGGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606368 MIM: 603901 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
APOA5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
APOA5 - apolipoprotein A5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166598.1 | 1048 | Silent Mutation | GCC,GCT | A,A 325 | NP_001160070.1 | |
NM_052968.4 | 1048 | Silent Mutation | GCC,GCT | A,A 325 | NP_443200.2 | |
XM_017017169.1 | 1048 | Silent Mutation | GCC,GCT | A,A 325 | XP_016872658.1 |
ZPR1 - ZPR1 zinc finger | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317086.1 | 1048 | Intron | NP_001304015.1 | |||
NM_003904.4 | 1048 | Intron | NP_003895.1 |