Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGCCTGGCTCACCCGGCCCCATC[C/T]GTCTCCGGTGGGCTCTTCTGCTGCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600644 | ||||||||||||||||||||
Literature Links: |
NECTIN1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NECTIN1 - nectin cell adhesion molecule 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002855.4 | 1312 | Intron | NP_002846.3 | |||
NM_203285.1 | 1312 | Silent Mutation | ACA,ACG | T,T 380 | NP_976030.1 | |
NM_203286.1 | 1312 | Intron | NP_976031.1 |