Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAACGGCATGAGAAGGAGAGACGCA[C/T]GTCCCAGGGACAGGAGAGCGCGCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606277 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TOLLIP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TOLLIP - toll interacting protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318512.1 | 1244 | UTR 3 | NP_001305441.1 | |||
NM_001318514.1 | 1244 | UTR 3 | NP_001305443.1 | |||
NM_001318515.1 | 1244 | UTR 3 | NP_001305444.1 | |||
NM_001318516.1 | 1244 | UTR 3 | NP_001305445.1 | |||
NM_019009.3 | 1244 | UTR 3 | NP_061882.2 | |||
XM_017017931.1 | 1244 | UTR 3 | XP_016873420.1 |