Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGGGGAAAGCCAGGGGGCACATAC[A/G]TCCCCATGCCGCCCCCTCCAAAGAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608541 | ||||||||||||||||||||
Literature Links: |
ARHGAP32 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ARHGAP32 - Rho GTPase activating protein 32 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142685.1 | 5318 | Missense Mutation | ACG,ATG | T,M 2047 | NP_001136157.1 | |
NM_014715.3 | 5318 | Missense Mutation | ACG,ATG | T,M 1698 | NP_055530.2 | |
XM_005271736.4 | 5318 | Missense Mutation | ACG,ATG | T,M 2061 | XP_005271793.2 | |
XM_011543072.2 | 5318 | Missense Mutation | ACG,ATG | T,M 2021 | XP_011541374.1 | |
XM_011543073.2 | 5318 | Missense Mutation | ACG,ATG | T,M 2034 | XP_011541375.2 | |
XM_011543075.2 | 5318 | Missense Mutation | ACG,ATG | T,M 1722 | XP_011541377.1 | |
XM_017018595.1 | 5318 | Missense Mutation | ACG,ATG | T,M 2021 | XP_016874084.1 | |
XM_017018596.1 | 5318 | Missense Mutation | ACG,ATG | T,M 1968 | XP_016874085.1 | |
XM_017018597.1 | 5318 | Intron | XP_016874086.1 |