Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATGGAGGACACTGTGCTTGTGGGC[A/G]TGGGCACACATGTGACTTCCATCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611142 MIM: 176930 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CKAP5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CKAP5 - cytoskeleton associated protein 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001008938.3 | 5867 | Missense Mutation | ACG,ATG | T,M 1914 | NP_001008938.1 | |
NM_014756.3 | 5867 | Missense Mutation | ACG,ATG | T,M 1854 | NP_055571.2 |
F2 - coagulation factor II, thrombin | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR5582 - microRNA 5582 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |