Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAGGCTGAGGTGCGCGTGTATGGG[A/G]GTGAGATCGTGCTGGCCCTGGAACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611205 MIM: 603606 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC88B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCDC88B - coiled-coil domain containing 88B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR1237 - microRNA 1237 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RPS6KA4 - ribosomal protein S6 kinase A4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001006944.1 | 490 | Missense Mutation | AGT,GGT | S,G 143 | NP_001006945.1 | |
NM_001300802.1 | 490 | Missense Mutation | AGT,GGT | S,G 143 | NP_001287731.1 | |
NM_001318361.1 | 490 | Missense Mutation | AGT,GGT | S,G 80 | NP_001305290.1 | |
NM_003942.2 | 490 | Missense Mutation | AGT,GGT | S,G 143 | NP_003933.1 | |
XM_005274380.1 | 490 | Missense Mutation | AGT,GGT | S,G 143 | XP_005274437.1 | |
XM_017018527.1 | 490 | Intron | XP_016874016.1 |