Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCTCCCCCTCTCCCCTCGCAGTGA[A/C]CCAACAGGAGAAGACCCCACCGCCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
C11orf84 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C11orf84 - chromosome 11 open reading frame 84 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_138471.2 | 444 | Missense Mutation | AAC,ACC | N,T 44 | NP_612480.1 | |
XM_005273782.2 | 444 | Missense Mutation | AAC,ACC | N,T 44 | XP_005273839.1 | |
XM_005273783.2 | 444 | Missense Mutation | AAC,ACC | N,T 44 | XP_005273840.1 | |
XM_006718437.1 | 444 | Missense Mutation | AAC,ACC | N,T 44 | XP_006718500.1 | |
XM_011544770.1 | 444 | Missense Mutation | AAC,ACC | N,T 44 | XP_011543072.1 | |
XM_011544771.1 | 444 | Missense Mutation | AAC,ACC | N,T 44 | XP_011543073.1 | |
XM_011544772.1 | 444 | Missense Mutation | AAC,ACC | N,T 44 | XP_011543074.1 |