Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCAGGAACCCGTAGATCTCCTCGG[A/T]CTTCTTCATAGGGTTCCGAGCCAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603797 | ||||||||||||||||||||
Literature Links: |
CHST1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CHST1 - carbohydrate sulfotransferase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003654.5 | 1607 | Missense Mutation | ACC,TCC | T,S 313 | NP_003645.1 | |
XM_006718356.3 | 1607 | Missense Mutation | ACC,TCC | T,S 313 | XP_006718419.1 | |
XM_017018459.1 | 1607 | Missense Mutation | ACC,TCC | T,S 313 | XP_016873948.1 |