Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTTCAGGCTGGAGGGCATTTTCCC[A/G]GCGGCGCTGCTGCCCCTGTTGCTGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616109 MIM: 608786 MIM: 605385 | ||||||||||||||||||||
Literature Links: |
C11orf80 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C11orf80 - chromosome 11 open reading frame 80 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001302084.1 | 382 | Intron | NP_001289013.1 | |||
NM_024650.3 | 382 | Intron | NP_078926.3 |
PC - pyruvate carboxylase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RCE1 - Ras converting CAAX endopeptidase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001032279.1 | 382 | Silent Mutation | CCA,CCG | P,P 10 | NP_001027450.1 | |
NM_005133.2 | 382 | Silent Mutation | CCA,CCG | P,P 114 | NP_005124.1 |