Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAAGCAGATGAGCGGGGAAGGGGG[C/T]GGCTCGGTCGGGCAGGGCGCGGGCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603940 MIM: 600050 | ||||||||||||||||||||
Literature Links: |
EHBP1L1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
EHBP1L1 - EH domain binding protein 1 like 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KCNK7 - potassium two pore domain channel subfamily K member 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MAP3K11 - mitogen-activated protein kinase kinase kinase 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002419.3 | 2563 | Silent Mutation | CCA,CCG | P,P 690 | NP_002410.1 |