Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCCCCTAGCATCTCCACTCCTCCA[A/G]TTGTTCCTGATATACAAAAGAATCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608876 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PCF11 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PCF11 - PCF11 cleavage and polyadenylation factor subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015885.3 | 913 | Missense Mutation | ATT,GTT | I,V 190 | NP_056969.2 | |
XM_005274048.1 | 913 | Missense Mutation | ATT,GTT | I,V 190 | XP_005274105.1 | |
XM_005274049.1 | 913 | Missense Mutation | ATT,GTT | I,V 190 | XP_005274106.1 | |
XM_017017918.1 | 913 | Missense Mutation | ATT,GTT | I,V 190 | XP_016873407.1 |