Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCACTGAGCAAACTGCAGTGCTTTT[C/T]TCCTCTGATCCTCAAAGCTTTCTCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
CWF19L2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CWF19L2 - CWF19-like 2, cell cycle control (S. pombe) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152434.2 | 2651 | Missense Mutation | AAA,AGA | K,R 874 | NP_689647.2 | |
XM_011542620.2 | 2651 | Intron | XP_011540922.1 | |||
XM_017017239.1 | 2651 | Missense Mutation | AAA,AGA | K,R 398 | XP_016872728.1 |
LOC105369477 - uncharacterized LOC105369477 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |