Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGTCTCTTTTCTTCTCTCTCTTTTA[A/G]TAAGGTGTTTCTGTCTACCAGTTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 123859 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CARS PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CARS - cysteinyl-tRNA synthetase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001014437.2 | 2049 | Silent Mutation | CTA,TTA | L,L 731 | NP_001014437.1 | |
NM_001194997.1 | 2049 | Silent Mutation | CTA,TTA | L,L 731 | NP_001181926.1 | |
NM_001751.5 | 2049 | Silent Mutation | CTA,TTA | L,L 648 | NP_001742.1 | |
NM_139273.3 | 2049 | Silent Mutation | CTA,TTA | L,L 648 | NP_644802.1 | |
XM_006718341.3 | 2049 | Silent Mutation | CTA,TTA | L,L 638 | XP_006718404.1 | |
XM_017018389.1 | 2049 | Silent Mutation | CTA,TTA | L,L 638 | XP_016873878.1 | |
XM_017018390.1 | 2049 | Silent Mutation | CTA,TTA | L,L 638 | XP_016873879.1 | |
XM_017018391.1 | 2049 | Silent Mutation | CTA,TTA | L,L 556 | XP_016873880.1 | |
XM_017018392.1 | 2049 | Silent Mutation | CTA,TTA | L,L 661 | XP_016873881.1 |