Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACCTGCACGAAGGAAGGCAGGCCC[A/G]GCCGGCAGCTGCTGGAGTCGGAGTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 143023 | ||||||||||||||||||||
Literature Links: |
LMNTD2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LMNTD2 - lamin tail domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173573.2 | 948 | Missense Mutation | NP_775844.2 | |||
XM_011519965.2 | 948 | Missense Mutation | XP_011518267.2 | |||
XM_011519967.2 | 948 | Missense Mutation | XP_011518269.2 | |||
XM_017017476.1 | 948 | Missense Mutation | XP_016872965.1 | |||
XM_017017477.1 | 948 | Missense Mutation | XP_016872966.1 | |||
XM_017017478.1 | 948 | Missense Mutation | XP_016872967.1 | |||
XM_017017479.1 | 948 | Missense Mutation | XP_016872968.1 |
LRRC56 - leucine rich repeat containing 56 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR210HG - MIR210 host gene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RASSF7 - Ras association domain family member 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |