Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGACCTCCTCCTTACCCTCTTACC[C/T]GGTAGTTACGAACACCATCCCAGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604773 MIM: 613739 MIM: 610027 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACAD8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ACAD8 - acyl-CoA dehydrogenase family member 8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
THYN1 - thymocyte nuclear protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001037304.1 | 392 | Missense Mutation | CAG,CGG | Q,R 97 | NP_001032381.1 | |
NM_001037305.1 | 392 | Missense Mutation | CAG,CGG | Q,R 97 | NP_001032382.1 | |
NM_014174.2 | 392 | Missense Mutation | CAG,CGG | Q,R 97 | NP_054893.1 | |
NM_199297.1 | 392 | Missense Mutation | CAG,CGG | Q,R 97 | NP_954994.1 | |
NM_199298.1 | 392 | Missense Mutation | CAG,CGG | Q,R 97 | NP_954995.1 | |
XM_005271527.3 | 392 | Missense Mutation | CAG,CGG | Q,R 138 | XP_005271584.1 | |
XM_005271528.3 | 392 | Missense Mutation | CAG,CGG | Q,R 138 | XP_005271585.1 |
VPS26B - VPS26, retromer complex component B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |