Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGAAATACTCCTCAGAAAAAAGCC[A/G]TCTTTGGGCAGTGTCGGGGTCTGCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611740 MIM: 605791 | ||||||||||||||||||||
Literature Links: |
BCO2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BCO2 - beta-carotene oxygenase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001037290.2 | 247 | Missense Mutation | ATC,GTC | I,V 10 | NP_001032367.2 | |
NM_001256397.1 | 247 | Missense Mutation | ATC,GTC | I,V 10 | NP_001243326.1 | |
NM_001256398.1 | 247 | Missense Mutation | ATC,GTC | I,V 44 | NP_001243327.1 | |
NM_001256400.1 | 247 | Intron | NP_001243329.1 | |||
NM_031938.5 | 247 | Missense Mutation | ATC,GTC | I,V 44 | NP_114144.4 |
TEX12 - testis expressed 12 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |