Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTACTGGTACTCAAAGTCTTTGG[A/G]TAGGCAATTCTGATGAAGATGAGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600814 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANKRD49 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ANKRD49 - ankyrin repeat domain 49 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017704.2 | 320 | Missense Mutation | ATA,GTA | I,V 46 | NP_060174.2 | |
XM_017017941.1 | 320 | Missense Mutation | ATA,GTA | I,V 46 | XP_016873430.1 |
MRE11A - MRE11 homolog A, double strand break repair nuclease | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005590.3 | 320 | Intron | NP_005581.2 | |||
NM_005591.3 | 320 | Intron | NP_005582.1 | |||
XM_005274008.3 | 320 | Intron | XP_005274065.1 | |||
XM_006718842.3 | 320 | Intron | XP_006718905.1 | |||
XM_011542837.2 | 320 | Intron | XP_011541139.1 | |||
XM_017017772.1 | 320 | Intron | XP_016873261.1 | |||
XM_017017773.1 | 320 | Intron | XP_016873262.1 |