Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCAGTCTGGAGGCCTTAGACAACC[C/T]GGCCAGGGGGTACGGTCTGTATGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
PPP1R32 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PPP1R32 - protein phosphatase 1 regulatory subunit 32 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001170753.1 | 350 | Missense Mutation | CCG,CTG | P,L 73 | NP_001164224.1 | |
NM_145017.2 | 350 | Missense Mutation | CCG,CTG | P,L 73 | NP_659454.2 |