Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTGGAACACCAGAAGCATGTGGG[G/T]GTGGCTCTAAGGGTCCTTTTGGAGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609568 | ||||||||||||||||||||
Literature Links: |
ARHGAP20 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ARHGAP20 - Rho GTPase activating protein 20 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001258415.1 | 3327 | Missense Mutation | CAC,CCC | H,P 1051 | NP_001245344.1 | |
NM_001258416.1 | 3327 | Missense Mutation | CAC,CCC | H,P 1048 | NP_001245345.1 | |
NM_001258417.1 | 3327 | Missense Mutation | CAC,CCC | H,P 1038 | NP_001245346.1 | |
NM_001258418.1 | 3327 | Missense Mutation | CAC,CCC | H,P 1038 | NP_001245347.1 | |
NM_020809.3 | 3327 | Missense Mutation | CAC,CCC | H,P 1074 | NP_065860.2 | |
XM_005271628.3 | 3327 | Missense Mutation | CAC,CCC | H,P 617 | XP_005271685.1 | |
XM_006718893.3 | 3327 | Missense Mutation | CAC,CCC | H,P 1074 | XP_006718956.1 |