Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGACGTTGGGGCTGAGGCTGGGG[A/G]ATAGAAGCCTTGCAGTGGTCCTCCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602013 | ||||||||||||||||||||
Literature Links: |
POLR2G PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
POLR2G - RNA polymerase II subunit G | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZBTB3 - zinc finger and BTB domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024784.3 | 911 | Intron | NP_079060.1 | |||
XM_011545256.2 | 911 | Missense Mutation | CCC,TCC | P,S 273 | XP_011543558.1 |