Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACCCAAGAGCGCCTCCAGTGGCAG[A/G]CGTGCTTTTGCCTCTGGATAAATGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608853 MIM: 615965 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BLID PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BLID - BH3-like motif containing, cell death inducer | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001786.2 | 425 | Silent Mutation | CGC,CGT | R,R 44 | NP_001001786.2 |
MIR100HG - mir-100-let-7a-2 cluster host gene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |