Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATTATTCCAGGTGAATGAGCCATG[C/T]GGAAATGTGGATTTATTAGGGTTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609673 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PDGFD PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PDGFD - platelet derived growth factor D | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_025208.4 | 3780 | UTR 3 | NP_079484.1 | |||
NM_033135.3 | 3780 | UTR 3 | NP_149126.1 |