Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCTGTGTCATCCAGGGAGCCAGAG[C/T]TGGCCTGGATGGACACCGTGTCTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604270 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LRP4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LRP4 - LDL receptor related protein 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002334.3 | 4871 | Missense Mutation | AAC,AGC | N,S 1853 | NP_002325.2 | |
XM_011520103.2 | 4871 | Missense Mutation | AAC,AGC | N,S 1585 | XP_011518405.1 | |
XM_011520104.2 | 4871 | Missense Mutation | AAC,AGC | N,S 1108 | XP_011518406.1 | |
XM_017017734.1 | 4871 | Intron | XP_016873223.1 |
LRP4-AS1 - LRP4 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |