Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATTCAGTGACCTTAGCTCCTCCAG[C/T]AAAACCGGTGTTAAACAATCTCTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
C11orf63 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C11orf63 - chromosome 11 open reading frame 63 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024806.3 | Intron | NP_079082.2 | ||||
NM_199124.2 | Intron | NP_954575.1 | ||||
XM_005271680.4 | Intron | XP_005271737.1 | ||||
XM_006718915.2 | Intron | XP_006718978.1 | ||||
XM_011543009.2 | Intron | XP_011541311.1 | ||||
XM_017018338.1 | Intron | XP_016873827.1 | ||||
XM_017018339.1 | Intron | XP_016873828.1 | ||||
XM_017018340.1 | Intron | XP_016873829.1 | ||||
XM_017018341.1 | Intron | XP_016873830.1 | ||||
XM_017018342.1 | Intron | XP_016873831.1 | ||||
XM_017018343.1 | Intron | XP_016873832.1 | ||||
XM_017018344.1 | Intron | XP_016873833.1 |