Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCCCACAGGCCACGTTCCACTACC[A/G]GACGCTGCACAGTGACGACGAGGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605555 MIM: 608794 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AIP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AIP - aryl hydrocarbon receptor interacting protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001302959.1 | 246 | UTR 5 | NP_001289888.1 | |||
NM_001302960.1 | 246 | Missense Mutation | CAG,CGG | Q,R 39 | NP_001289889.1 | |
NM_003977.3 | 246 | Missense Mutation | CAG,CGG | Q,R 39 | NP_003968.3 |
MIR6752 - microRNA 6752 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PITPNM1 - phosphatidylinositol transfer protein membrane associated 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |