Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGGTGAGATGGTGGTTGAGGTGTC[C/T]GGCGAGGATTTACGTGTATTATTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609201 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
UBASH3B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
UBASH3B - ubiquitin associated and SH3 domain containing B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032873.4 | Intron | NP_116262.2 | ||||
XM_005271712.3 | Intron | XP_005271769.1 | ||||
XM_011543041.2 | Intron | XP_011541343.1 | ||||
XM_011543042.1 | Intron | XP_011541344.1 |