Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCGTTCTCCGTGTTGTCATTCCCC[A/G]AGGAGCTCACGAGACACTCCTTCCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608256 | ||||||||||||||||||||
Literature Links: |
SCN4B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SCN4B - sodium voltage-gated channel beta subunit 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142348.1 | 457 | Missense Mutation | TCG,TTG | S,L 72 | NP_001135820.1 | |
NM_001142349.1 | 457 | Missense Mutation | TCG,TTG | S,L 96 | NP_001135821.1 | |
NM_174934.3 | 457 | Missense Mutation | TCG,TTG | S,L 206 | NP_777594.1 |