Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTCTGGTCTGTGTAGCCACCATGT[A/G]CAGTGCCCAGGACAGGACTGATCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 136425 MIM: 600829 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FOLR2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FOLR2 - folate receptor beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000803.4 | 213 | Missense Mutation | TAC,TGC | Y,C 19 | NP_000794.3 | |
NM_001113534.1 | 213 | Missense Mutation | TAC,TGC | Y,C 19 | NP_001107006.1 | |
NM_001113535.1 | 213 | Missense Mutation | TAC,TGC | Y,C 19 | NP_001107007.1 | |
NM_001113536.1 | 213 | Missense Mutation | TAC,TGC | Y,C 19 | NP_001107008.1 | |
XM_005273856.3 | 213 | Missense Mutation | TAC,TGC | Y,C 28 | XP_005273913.1 |
INPPL1 - inositol polyphosphate phosphatase like 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |