Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGACTGTGATGAAAAGGGTCACCCC[A/G]AGTAACAGCACCACCAGAAGGAGAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610880 | ||||||||||||||||||||
Literature Links: |
C11orf24 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C11orf24 - chromosome 11 open reading frame 24 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300913.1 | 1666 | Intron | NP_001287842.1 | |||
NM_022338.3 | 1666 | Silent Mutation | CTC,CTT | L,L 408 | NP_071733.1 | |
XM_005274053.3 | 1666 | Silent Mutation | CTC,CTT | L,L 408 | XP_005274110.1 | |
XM_011545103.2 | 1666 | Silent Mutation | CTC,CTT | L,L 376 | XP_011543405.1 |