Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGACCCTGCGCACGCTGCCCGGCAC[A/G]CGGCTCGCCTGGCTGGCGGAGCCCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 176258 | ||||||||||||||||||||
Literature Links: |
KCNC1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KCNC1 - potassium voltage-gated channel subfamily C member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001112741.1 | 151 | Silent Mutation | ACA,ACG | T,T 32 | NP_001106212.1 | |
NM_004976.4 | 151 | Silent Mutation | ACA,ACG | T,T 32 | NP_004967.1 | |
XM_011520080.2 | 151 | Silent Mutation | ACA,ACG | T,T 32 | XP_011518382.1 |