Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGCGGTGGGGCTGCCCCTGCTGCT[C/T]ATCTCGCTGGCCTTCGCGCAGGAGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608420 | ||||||||||||||||||||
Literature Links: |
PANX1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PANX1 - pannexin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015368.3 | 335 | Silent Mutation | CTC,CTT | L,L 49 | NP_056183.2 | |
XM_011542734.2 | 335 | Intron | XP_011541036.1 | |||
XM_017017464.1 | 335 | UTR 5 | XP_016872953.1 |