Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTCGGGGCCCACTTTGCTGTCATC[A/C]GCCGAGCGTCCCTGGAGAAGAACCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611033 MIM: 603853 | ||||||||||||||||||||
Literature Links: |
C11orf21 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C11orf21 - chromosome 11 open reading frame 21 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_011520034.1 | 261 | Intron | XP_011518336.1 |
TSPAN32 - tetraspanin 32 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_139022.2 | 261 | Missense Mutation | AGC,CGC | S,R 45 | NP_620591.3 | |
XM_005252719.4 | 261 | Missense Mutation | AGC,CGC | S,R 80 | XP_005252776.1 | |
XM_005252720.4 | 261 | Missense Mutation | AGC,CGC | S,R 80 | XP_005252777.1 | |
XM_011519815.2 | 261 | Missense Mutation | AGC,CGC | S,R 80 | XP_011518117.2 | |
XM_011519818.2 | 261 | Missense Mutation | AGC,CGC | S,R 80 | XP_011518120.2 | |
XM_011519819.1 | 261 | Missense Mutation | AGC,CGC | S,R 15 | XP_011518121.1 | |
XM_011519822.1 | 261 | Missense Mutation | AGC,CGC | S,R 80 | XP_011518124.1 | |
XM_011519828.2 | 261 | Missense Mutation | AGC,CGC | S,R 45 | XP_011518130.1 | |
XM_011519829.2 | 261 | Missense Mutation | AGC,CGC | S,R 80 | XP_011518131.2 | |
XM_017017064.1 | 261 | Intron | XP_016872553.1 | |||
XM_017017065.1 | 261 | Intron | XP_016872554.1 | |||
XM_017017066.1 | 261 | Intron | XP_016872555.1 | |||
XM_017017067.1 | 261 | Missense Mutation | AGC,CGC | S,R 15 | XP_016872556.1 | |
XM_017017068.1 | 261 | Intron | XP_016872557.1 | |||
XM_017017069.1 | 261 | Intron | XP_016872558.1 | |||
XM_017017070.1 | 261 | Intron | XP_016872559.1 | |||
XM_017017071.1 | 261 | Intron | XP_016872560.1 |